missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexyl N-(4-Aminobenzenecarboximidoyl)carbamate Hydrochloride, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Hexyl N-(4-Aminobenzenecarboximidoyl)carbamate Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Hexyl N-(4-Aminobenzenecarboximidoyl)carbamate Hydrochloride |
| CAS | 1307233-93-7 |
| Analyte or Component Names | Hexyl N-(4-Aminobenzenecarboximidoyl)carbamate Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H21 N3 O2 . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Hexyl N-(4-aminobenzenecarboximidoyl)carbamate hydrochloride, N-[(4-aminophenyl)iminomethyl]-Carbamic acid hexyl ester hydrochloride (1:1) |
| InChI Formula | InChI=1S/C14H21N3O2.ClH/c1-2-3-4-5-10-19-14(18)17-13(16)11-6-8-12(15)9-7-11;/h6-9H,2-5,10,15H2,1H3,(H2,16,17,18);1H |
| SMILES | Cl.CCCCCCOC(=O)NC(=N)c1ccc(N)cc1 |
| IUPAC Name | hexyl N-(4-aminobenzenecarboximidoyl)carbamate;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction