missing translation for 'onlineSavingsMsg'
Learn More

Hexyl acetate, 99%

Product Code. 10103390 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
25mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10103390 1 L Glass bottle 1 L
10593691 25 mL Glass bottle 25mL
11984911 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10103390 Supplier Thermo Scientific Acros Supplier No. 148500010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 142-92-7 | C8H16O2 | 144.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-92-7
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.21
InChI Key AOGQPLXWSUTHQB-UHFFFAOYSA-N
Synonym n-hexyl acetate, 1-hexyl acetate, hexyl ethanoate, acetic acid hexyl ester, n-hexyl ethanoate, acetic acid, hexyl ester, hexyl alcohol, acetate, l-hexyl acetate, fema no. 2565, natural
PubChem CID 8908
ChEBI CHEBI:87510
IUPAC Name hexyl acetate
SMILES CCCCCCOC(=O)C

Specifications

Acidity 1mg KOH/g max.
Melting Point -80.0°C
Color Colorless to Yellow
Density 0.8710g/mL
Boiling Point 168.0°C to 170.0°C
Flash Point 43°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CO2C6H13
Refractive Index 1.4070 to 1.4110
Quantity 1 L
Beilstein 02,132
Specific Gravity 0.871
Solubility Information Solubility in water: immiscible. Other solubilities: soluble in alcohol and ether
Formula Weight 144.21
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Hexyl acetate
Show More Show Less
compliance-icons
Product Identifier
  • Hexyl acetate
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.