missing translation for 'onlineSavingsMsg'
Learn More

Hexosaminidase A/HEXA Antibody (3F10) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

£161.00 - £435.00

Specifications

Antigen Hexosaminidase A/HEXA
Clone 3F10
Concentration 1 mg/mL
Dilution Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready
Applications Western Blot, Flow Cytometry, ELISA, CyTOF
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18191089
View Documents
Novus Biologicals
NBP2-37505
0.1 mL
£435.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18636505
View Documents
Novus Biologicals
NBP2-37505-0.025ml
0.025 mL
£161.00
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Hexosaminidase A/HEXA Monoclonal specifically detects Hexosaminidase A/HEXA in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.
Specifications

Specifications

Hexosaminidase A/HEXA
1 mg/mL
Western Blot, Flow Cytometry, ELISA, CyTOF
Unconjugated
RUO
Human
beta-hexosaminidase subunit alpha, Beta-N-acetylhexosaminidase subunit alpha, EC 3.2.1, EC 3.2.1.52, hexosaminidase A (alpha polypeptide), Hexosaminidase subunit A, MGC99608, N-acetyl-beta-glucosaminidase subunit alpha, TSD
HEXA
IgG2b
Affinity Purified
3F10
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready
Monoclonal
Mouse
Neuroscience, Signal Transduction
PBS with 0.05% Sodium Azide
3073
Purified recombinant fragment of human HEXA expressed in E. Coli.
Primary
60.7 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.