missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Hexokinase Recombinant Rabbit Monoclonal Antibody (HL2000)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556736
This item is not returnable.
View return policy
Description
Hexokinase Recombinant Monoclonal Antibody for Western Blot
Hexokinases phosphorylate glucose to produce glucose-6-phosphate, the first step in most glucose metabolism pathways. This gene encodes a ubiquitous form of hexokinase which localizes to the outer membrane of mitochondria. Mutations in this gene have been associated with hemolytic anemia due to hexokinase deficiency. Alternative splicing of this gene results in five transcript variants which encode different isoforms, some of which are tissue-specific. Each isoform has a distinct N-terminus; the remainder of the protein is identical among all the isoforms. A sixth transcript variant has been described, but due to the presence of several stop codons, it is not thought to encode a protein.
Specifications
| Hexokinase | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Rabbit | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| HL2000 | |
| Unconjugated | |
| hexokinase 1; Hexokinase PI; Hexokinase-1; Hexokinase-A; HK1-ta; HK1-tb; HK1-tc; HKA; HKI; HXK1; YFR053C | |
| Recombinant fragment of human Hexokinase II. | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction