missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexokinase 2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22029-1-AP-20UL
This item is not returnable.
View return policy
Description
In vertebrates there are four major glucose-phosphorylating isoenzymes, designated hexokinase I, II, III, and IV. Hexokinase activity is involved in the first step in several metabolic pathways including phosphorylation of glucose to produce glucose-6-phosphate, thus committing glucose to the glycolytic pathway. Hexokinase is an allosteric enzyme inhibited by its product GLC-6-P. Hexokinase 2 is the predomit hexokinase isozyme expressed in insulin-responsive tissues such as skeletal muscle. Expression of this gene is insulin-responsive, and studies in rat suggest that it is involved in the increased rate of glycolysis seen in rapidly growing cancer cells.Specifications
| Hexokinase 2 | |
| Polyclonal | |
| Unconjugated | |
| HK2 | |
| Hexokinase 2, Hexokinase type II, HK II, HK2, HKII, HXK2, Muscle form hexokinase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 15277, 25059, 3099 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.34 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O08528, P27881, P52789 | |
| Hk2 | |
| Hexokinase 2 Fusion Protein Ag16895 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction