missing translation for 'onlineSavingsMsg'
Learn More

Hexokinase 2 Antibody (4H1), Novus Biologicals™

Product Code. 18370028 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18370028 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18370028 Supplier Novus Biologicals Supplier No. H00003099M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Hexokinase 2 Monoclonal antibody specifically detects Hexokinase 2 in Human, Rat samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hexokinase 2
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 4H1
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. P52789
Gene Alias DKFZp686M1669, EC 2.7.1, hexokinase 2, Hexokinase type II, HKII, HXK2, muscle, Muscle form hexokinase
Host Species Mouse
Immunogen HK2 (AAH21116, 818 a.a. ~ 917 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. IVKEVCTVVARRAAQLCGAGMAAVVDRIRENRGLDALKVTVGVDGTLYKLHPHFAKVMHETVKDLAPKCDVSFLQSEDGSGKGAALITAVACRIREAGQR
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 3099
Target Species Human, Rat
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.