missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexokinase 2 Antibody (1A7), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-04296AF700
This item is not returnable.
View return policy
Description
Hexokinase 2 Monoclonal specifically detects Hexokinase 2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Hexokinase 2 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| HK2 | |
| Recombinant human Hexokinase 2 (1-917aa) purified from E. coli | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 3099 | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 1A7 | |
| Western Blot, ELISA | |
| DKFZp686M1669, EC 2.7.1, hexokinase 2, Hexokinase type II, HKII, HXK2, muscle, Muscle form hexokinase | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Anti-human Hexokinase 2 mAb, clone 1A7, is derived from hybridization of mouse FO myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human Hexokinase 2 protein. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction