missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexokinase 1 Antibody (4D7), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-04295AF594
This item is not returnable.
View return policy
Description
Hexokinase 1 Monoclonal antibody specifically detects Hexokinase 1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| Hexokinase 1 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| Brain form hexokinase, EC 2.7.1, EC 2.7.1.1, glycolytic enzyme, hexokinase 1, Hexokinase type I, hexokinase-1, HK I, HK1-ta, HK1-tb, HK1-tc, HKI, HXK1 | |
| Recombinant human Hexokinase1 (1-917aa) purified from E. coli | |
| 0.1 mL | |
| Cancer | |
| 3098 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| 4D7 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction