missing translation for 'onlineSavingsMsg'
Learn More

Hexocyclium Metilsulfate, LoGiCal

Product Code. 30457429
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457429 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457429 Supplier LoGiCal Supplier No. LGCFOR1655.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hexocyclium Metilsulfate
CAS 115-63-9
Analyte or Component Names Hexocyclium Metilsulfate
Shipping Condition Room Temperature
Molecular Formula C20 H33 N2 O . C H3 O4 S
Certifications/Compliance ISO 9001
Synonym 4-(β-Cyclohexyl-β-hydroxyphenethyl)-1,1-dimethylpiperazinium methylsulfate, Piperazinium, 4-(2-cyclohexyl-2-hydroxy-2-phenylethyl)-1,1-dimethyl-, methyl sulfate (salt), Piperazinium, 4-(β-cyclohexyl-β-hydroxyphenethyl)-1,1-dimethyl-, methyl sulfate, (±)-Hexocyclium methyl sulfate, 4-(β-Cyclohexyl-β-hydroxy-β-phenethyl)-1,1-dimethylpiperazinum methyl sulfate, AMA-DE 1973, Hexocyclium, Hexocyclium methosulfate, Hexocyclium methyl sulfate, Hexocyclium metilsulfate, MI Tral, N-(β-Cyclohexyl-β-hydroxy-β-phenylethyl)-N'-methylpiperazine dimethylsulfate, N-(β-Cyclohexyl-β-hydroxy-β-phenylethyl)-N'-methylpiperazine methosulfate, NSC 30256, Tral, Tralin, Traline
InChI Formula InChI=1S/C20H33N2O.CH4O4S/c1-22(2)15-13-21(14-16-22)17-20(23,18-9-5-3-6-10-18)19-11-7-4-8-12-19;1-5-6(2,3)4/h3,5-6,9-10,19,23H,4,7-8,11-17H2,1-2H3;1H3,(H,2,3,4)/q+1;/p-1
Solution Type Neat
SMILES COS(=O)(=O)[O-].C[N+]1(C)CCN(CC(O)(C2CCCCC2)c3ccccc3)CC1
IUPAC Name 1-cyclohexyl-2-(4,4-dimethylpiperazin-4-ium-1-yl)-1-phenylethanol;methyl sulfate
Industry Type Clinical Testing
Molecular Weight (g/mol) 428.59
Quantity 10 mg
Formula Weight 428.2345 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.