missing translation for 'onlineSavingsMsg'
Learn More

Hexobendine Dihydrochloride, TRC

Product Code. 30394804
Change view
Click to view available options
Quantity:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30394804 250 mg
30389133 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30394804 Supplier Toronto Research Chemicals Supplier No. TRCH295450250MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-62-4
Molecular Formula C30 H44 N2 O10 . 2 Cl H
Molecular Weight (g/mol) 665.6
Synonym 3,4,5-Trimethoxybenzoic Acid 1,2-Ethanediylbis[(methylimino)-3,1-propanediyl] Ester Dihydrochloride, 3,3'-[Ethylenebis(methylimino)]di-1-propanol Bis(3,4,5-trimethoxybenzoate) (Ester) Dihydrochloride, Andiamine, Hexobendin Hydrochloride, Reoxyl
IUPAC Name 3-[methyl-[2-[methyl-[3-(3,4,5-trimethoxybenzoyl)oxypropyl]amino]ethyl]amino]propyl 3,4,5-trimethoxybenzoate;dihydrochloride
SMILES Cl.Cl.COc1cc(cc(OC)c1OC)C(=O)OCCCN(C)CCN(C)CCCOC(=O)c2cc(OC)c(OC)c(OC)c2

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C30H44N2O10.2ClH/c1-31(11-9-15-41-29(33)21-17-23(35-3)27(39-7)24(18-21)36-4)13-14-32(2)12-10-16-42-30(34)22-19-25(37-5)28(40-8)26(20-22)38-6;;/h17-20H,9-16H2,1-8H3;2*1H
Formula Weight 664.253
Chemical Name or Material Hexobendine Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.