missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEXO Antibody (OTI1B3), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72312AF350
This item is not returnable.
View return policy
Description
HEXO Monoclonal specifically detects HEXO in Human, Rat, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| HEXO | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| ERI1 | |
| Full length recombinant protein of human HEXO (NP_699163) produced in HEK293T cells. | |
| Primary | |
| Human, Rat, Primate, Monkey | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| OTI1B3 | |
| Western Blot, Immunohistochemistry | |
| 3′ exoribonuclease, 3′-5′ exonuclease ERI1, 3′EXO, 3′HEXO, EC 3.1, enhanced RNAi three prime mRNA exonuclease homolog 1, Eri-1 homolog, exoribonuclease 1, HEXO, histone mRNA 3′ end-specific exonuclease, Histone mRNA 3′-end-specific exoribonuclease, Histone mRNA 3′-exonuclease 1, MGC35395, Protein 3′hExo, THEX1, three prime histone mRNA exonuclease 1,3′-5′ exoribonuclease 1, three prime mRNA exonuclease 1 | |
| Mouse | |
| 0.1 mL | |
| 90459 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering