missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEXIM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15676-1-AP-150UL
This item is not returnable.
View return policy
Description
15676-1-AP recognizes both 70 and 54 kDa bands.
The protein encoded by this gene is a member of the RecQ DNA helicase family. DNA helicases are enzymes involved in various types of DNA repair, including mismatch repair, nucleotide excision repair and direct repair. Some members of this family are associated with genetic disorders with predisposition to maligcy and chromosomal instability. The biological function of this helicase has not yet been determined. Two alternatively spliced transcripts, which encode the same isoform but differ in their 5′ and 3′ UTRs, have been described.Specifications
| HEXIM1 | |
| Polyclonal | |
| Unconjugated | |
| HEXIM1 | |
| Cardiac lineage protein 1, CLP1, EDG1, HEXIM1, HIS1, MAQ1, Menage a quatre protein 1, Protein HEXIM1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10614, 192231, 498008 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O94992, Q5M9G1, Q8R409 | |
| Hexim1 | |
| HEXIM1 Fusion Protein Ag8144 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction