missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEXIM1 Mouse anti-Human, Clone: 2C8A2, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | HEXIM1 |
|---|---|
| Clone | 2C8A2 |
| Concentration | 0.3 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16808075
|
Proteintech
66311-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16898065
|
Proteintech
66311-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
66311-1-Ig recognizes the 54 kDa band.
The protein encoded by this gene is a member of the RecQ DNA helicase family. DNA helicases are enzymes involved in various types of DNA repair, including mismatch repair, nucleotide excision repair and direct repair. Some members of this family are associated with genetic disorders with predisposition to maligcy and chromosomal instability. The biological function of this helicase has not yet been determined. Two alternatively spliced transcripts, which encode the same isoform but differ in their 5′ and 3′ UTRs, have been described.Specifications
| HEXIM1 | |
| 0.3 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Cardiac lineage protein 1, CLP1, EDG1, HEXIM1, HIS1, MAQ1, Menage a quatre protein 1, Protein HEXIM1 | |
| HEXIM1 | |
| IgM | |
| Affinity Chromatography | |
| Antibody |
| 2C8A2 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| O94992 | |
| 10614 | |
| HEXIM1 Fusion Protein Ag8019 | |
| Primary | |
| -20°C | |
| HEXIM1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title