missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HEXIM1 Monoclonal Antibody (K40046_16G4), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA555786
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: WGQQQRQLGK KKHRRRPSKK KRHWKPYYKL TWEEKKKFDE KQSLRASRIR AEMFAKGQPV APYNTTQFLM DDHDQEEPDL KTGLYSKRAA AKSDDTSDDD FMEEGGEEDG GSDGMGGDGS EFLQRDFSET YERYHTESLQ NMSKQELIKE YLELEKCLSR MEDENNRLRL ESKRLGGDDA RVRELELELD RLRAENLQLL
The protein encoded by this gene is a member of the RecQ DNA helicase family. DNA helicases are enzymes involved in various types of DNA repair, including mismatch repair, nucleotide excision repair and direct repair. Some members of this family are associated with genetic disorders with predisposition to malignancy and chromosomal instability. The biological function of this helicase has not yet been determined. Two alternatively spliced transcripts, which encode the same isoform but differ in their 5' and 3' UTRs, have been described.
Specifications
| HEXIM1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| O94992 | |
| HEXIM1 | |
| Recombinant human HEXIM1 (immunogen range is 141-340). | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| K40046_16G4 | |
| Unconjugated | |
| HEXIM1 | |
| 7330426E13Rik; Cardiac lineage protein 1; CLP1; Clp-1; EDG1; Estrogen down-regulated gene 1 protein; FLJ13562; hexamethylene bisacetamide inducible 1; hexamethylene bis-acetamide inducible 1; hexamethylene bisacetamide-inducible protein; hexamethylene bis-acetamide-inducible protein 1; hexamethylene-bis-acetamide-inducible transcript 1; hexamthylene bis-acetamide inducible 1; HEXIM1; HGNC:24953; HIS1; HMBA-inducible; MAQ1; menage a quatre 1; menage a quatre protein 1; Protein HEXIM1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 10614 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction