missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexim 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-71866
This item is not returnable.
View return policy
Description
Hexim 1 Polyclonal specifically detects Hexim 1 in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| Hexim 1 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| Cardiac lineage protein 1, CLP-1, CLP1hexamethylene-bis-acetamide-inducible transcript 1, EDG1HMBA-inducible, Estrogen down-regulated gene 1 protein, hexamethylene bis-acetamide inducible 1, Hexamethylene bis-acetamide-inducible protein 1, hexamthylene bis-acetamide inducible 1, HIS1FLJ13562, HMBA-inducible, MAQ1hexamethylene bisacetamide-inducible protein, menage a quatre 1, Menage a quatre protein 1, protein HEXIM1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Immunohistochemistry 1:500- 1:2000, Immunoprecipitation 2 - 10 ug/mg lysate, Immunohistochemistry-Paraffin 1:500 - 1:2000 | |
| NP_006451.1 | |
| HEXIM1 | |
| The immunogen for this product maps to a region between residue 1 and 50 of human Hexamethylene Bis-Acetamide Inducible 1 using the numbering given in entry NP_006451.1 (GeneID 10614). | |
| 100 μL | |
| Cell Cycle and Replication | |
| 10614 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction