missing translation for 'onlineSavingsMsg'
Learn More

Hexim 1 Antibody [mFluor Violet 450 SE], Novus Biologicals™

Product Code. 30735207 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30735207 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735207 Supplier Novus Biologicals Supplier No. AF8106MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Hexim 1 Polyclonal antibody specifically detects Hexim 1 in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hexim 1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Formulation 50mM Sodium Borate
Gene Alias Cardiac lineage protein 1, CLP-1, CLP1hexamethylene-bis-acetamide-inducible transcript 1, EDG1HMBA-inducible, Estrogen down-regulated gene 1 protein, hexamethylene bis-acetamide inducible 1, Hexamethylene bis-acetamide-inducible protein 1, hexamthylene bis-acetamide inducible 1, HIS1FLJ13562, HMBA-inducible, MAQ1hexamethylene bisacetamide-inducible protein, menage a quatre 1, Menage a quatre protein 1, protein HEXIM1
Host Species Sheep
Immunogen E. coli-derived recombinant human Hexim 1, His213-Asp359, Accession # O94992
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 10614
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.