missing translation for 'onlineSavingsMsg'
Learn More

Hexetidine, Mixture of Stereoisomers, TRC

Product Code. 30393843
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30393843 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30393843 Supplier Toronto Research Chemicals Supplier No. TRCH9250131G
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hexetidine
CAS 141-94-6
Molecular Formula C21 H45 N3
Quantity 1 g
Synonym Hexetidine, 1,3-bis(2-ethylhexyl)]-5-methylhexahydropyrimidin-5-amine
InChI Formula InChI=1S/C21H45N3/c1-6-10-12-19(8-3)14-23-16-21(5,22)17-24(18-23)15-20(9-4)13-11-7-2/h19-20H,6-18,22H2,1-5H3
SMILES CCCCC(CC)CN1CN(CC(CC)CCCC)CC(C)(N)C1
IUPAC Name 1,3-bis(2-ethylhexyl)-5-methyl-1,3-diazinan-5-amine
Molecular Weight (g/mol) 339.6
Formula Weight 339.3613
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.