missing translation for 'onlineSavingsMsg'
Learn More

Hexetidine, mixture of stereoisomers, 97%, Thermo Scientific™

Product Code. p-7047320
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11309475

Brand: Thermo Scientific Alfa Aesar L18699.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-94-6
Molecular Formula C21H45N3
Molecular Weight (g/mol) 339.612
MDL Number MFCD00010428
InChI Key DTOUUUZOYKYHEP-UHFFFAOYSA-N
Synonym hexetidine, oraldene, hexoral, glypesin, hextril, elsix, collu hextril, sterilate, sterisil, triocil
PubChem CID 3607
IUPAC Name 1,3-bis(2-ethylhexyl)-5-methyl-1,3-diazinan-5-amine
SMILES CCCCC(CC)CN1CC(CN(C1)CC(CC)CCCC)(C)N

Specifications

Density 0.889
Boiling Point 160°C (0.4 mmHg)
Flash Point 70°C (158°F)
Refractive Index 1.4645
Quantity 5 g
Beilstein 161071
Merck Index 14,4703
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 339.61
Percent Purity 97%
Chemical Name or Material Hexetidine, mixture of stereoisomers
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.