missing translation for 'onlineSavingsMsg'
Learn More

Hexetidine, MedChemExpress

Product Code. 16442299
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16442299 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16442299 Supplier Medchem Express Supplier No. HYB0996

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Hexetidine is an orally active antiseptic with broad antibacterial and antifungal activity. Hexetidine give important potential for treatment of oral infections.

Hexetidine is an orally active antiseptic with broad antibacterial and antifungal activity. Hexetidine give important potential for treatment of oral infections.

Specifications

Chemical Name or Material Hexetidine
Physical Form Liquid
CAS 141-94-6
Color Colorless
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C21H45N3
For Use With (Application) COVID-19-immunoregulation
Synonym NSC-17764
Solubility Information DMSO : ≥ 100 mg/mL (294.46 mM)
SMILES NC1(C)CN(CC(CC)CCCC)CN(CC(CC)CCCC)C1
Molecular Weight (g/mol) 339.6
Formula Weight 339.6
Percent Purity 97.0%
Grade Research
Afficher plus Afficher moins

Research purposes only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.