missing translation for 'onlineSavingsMsg'
Learn More

Hexestrol, TRC

Product Code. 30389132
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30389132 100 mg
30393597 500 mg
30401848 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30389132 Supplier Toronto Research Chemicals Supplier No. TRCH295300100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84-16-2
Molecular Formula C18 H22 O2
Molecular Weight (g/mol) 270.37
Synonym Phenol, 4,4'-(1,2-diethyl-1,2-ethanediyl)bis-, (R*,S*)-, Phenol, 4,4'-(1,2-diethylethylene)di-, meso- (8CI), Cycloestrol, Erythrohexestrol, Estrifar, Estronal, Extra-Plex, Hexanoestrol, Hexestrofen, Hexestrol, Hexoestrol, Hexron, Hormoestrol, Mesohexestrol, NSC 9894, Sinestrol, Synestrol, Synoestrol, Synthovo, Syntrogene, meso-3,4-Bis(p-hydroxyphenyl)-n-hexane, meso-3,4-Di(p-hydroxyphenyl)-n-hexane, meso-Hexestrol, Phenol, 4,4'-[(1R,2S)-1,2-diethyl-1,2-ethanediyl]bis-, rel-
IUPAC Name 4-[(3S,4R)-4-(4-hydroxyphenyl)hexan-3-yl]phenol
SMILES CC[C@H]([C@H](CC)c1ccc(O)cc1)c2ccc(O)cc2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C18H22O2/c1-3-17(13-5-9-15(19)10-6-13)18(4-2)14-7-11-16(20)12-8-14/h5-12,17-20H,3-4H2,1-2H3/t17-,18+
Formula Weight 270.162
Percent Purity >95%
Chemical Name or Material Hexestrol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.