missing translation for 'onlineSavingsMsg'
Learn More

Hexen-5-ol, TRC

Product Code. 30391401
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391401 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30391401 Supplier Toronto Research Chemicals Supplier No. TRCH29502550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hexen-5-ol
CAS 626-94-8
Molecular Formula C7 H4 Cl F3 O2
Quantity 50 mg
Synonym 5-Hydroxy-1-hexene
InChI Formula InChI=1S/C7H4ClF3O2/c8-5-3-4(12)1-2-6(5)13-7(9,10)11/h1-3,12H
SMILES Oc1ccc(OC(F)(F)F)c(Cl)c1
IUPAC Name 3-chloro-4-(trifluoromethoxy)phenol
Molecular Weight (g/mol) 212.554
Formula Weight 211.985
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.