missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexazinone metabolite A 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Hexazinone metabolite A |
| CAS | 72576-13-7 |
| Analyte or Component Names | Hexazinone metabolite A |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H20 N4 O3 |
| Concentration | 100 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | 1,3,5-Triazine-2,4(1H,3H)-dione, 6-(dimethylamino)-3-(4-hydroxycyclohexyl)-1-methyl-, 6-(Dimethylamino)-3-(4-hydroxycyclohexyl)-1-methyl-1,3,5-triazine-2,4(1H,3H)-dione, rel-trans-6-(Dimethylamino)-3-(4-hydroxycyclohexyl)-1-methyl-1,3,5-Triazine-2,4(1H,3H)-dione |
| InChI Formula | InChI=1S/C12H20N4O3/c1-14(2)10-13-11(18)16(12(19)15(10)3)8-4-6-9(17)7-5-8/h8-9,17H,4-7H2,1-3H3 |
| SMILES | CN(C)C1=NC(=O)N(C2CCC(O)CC2)C(=O)N1C |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?