missing translation for 'onlineSavingsMsg'
Learn More

Hexarhodium hexadecacarbonyl, 98%

Product Code. 11491217
Change view
Click to view available options
Quantity:
0.1 g
0.5 g
Unit Size:
0.1g
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11491217 0.5 g 0.5g
11481217 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11491217 Supplier Thermo Scientific Alfa Aesar Supplier No. B24185.02
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 28407-51-4 | C16O16Rh6 | 1065.593 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28407-51-4
Molecular Formula C16O16Rh6
Molecular Weight (g/mol) 1065.593
MDL Number MFCD00011206
InChI Key SZQABOJVTZVBHE-UHFFFAOYSA-N
Synonym hexarhodium hexadecacarbonyl, hexarhodiumhexadecacarbonyl, rhodium 0 carbonyl, rh6 co 16, hexarhodium 0 hexadecacarbonyl, hexadecacarbonylhexarhodium, hexarhodium 0 hexadecacarbonyl, rh 57-60 % approx.
PubChem CID 10866043
IUPAC Name carbon monoxide;rhodium
SMILES [C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Rh].[Rh].[Rh].[Rh].[Rh].[Rh]

Specifications

Melting Point >300°C
Quantity 0.5 g
UN Number UN3466
Formula Weight 1065.6
Percent Purity 98%
Chemical Name or Material Hexarhodium hexadecacarbonyl

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.