missing translation for 'onlineSavingsMsg'
Learn More

Hexaphenylbenzene, 98+%

Product Code. 11370044
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11370044

Brand: Thermo Scientific Alfa Aesar L00446.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 992-04-1 | C42H30 | 534.702 g/mol

Hexaphenylbenzene was used to prepare the fluorescent nanorods used for the detection of trinitrotoulene (TNT).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexaphenylbenzene was used to prepare the fluorescent nanorods used for the detection of trinitrotoulene (TNT).

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 992-04-1
Molecular Formula C42H30
Molecular Weight (g/mol) 534.702
MDL Number MFCD00003057
InChI Key QBHWPVJPWQGYDS-UHFFFAOYSA-N
Synonym hexaphenylbenzene, benzene, hexaphenyl, 3',4',5',6'-tetraphenyl-o-terphenyl, acmc-20akri, m-terphenyl, 2',4',5',6'-tetraphenyl, ghl.pd_mitscher_leg0.890, hexaphenylbenzene 1g, 1,1':2',1-terphenyl, 3',4',5',6'-tetraphenyl, 1,2,3,4,5,6-hexaphenylbenzene
PubChem CID 70432
IUPAC Name 1,2,3,4,5,6-hexakis-phenylbenzene
SMILES C1=CC=C(C=C1)C2=C(C(=C(C(=C2C3=CC=CC=C3)C4=CC=CC=C4)C5=CC=CC=C5)C6=CC=CC=C6)C7=CC=CC=C7

Specifications

Melting Point >300°C
Quantity 5 g
Beilstein 1895903
Solubility Information Insoluble in water.
Formula Weight 534.7
Percent Purity ≥98%
Chemical Name or Material Hexaphenylbenzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.