missing translation for 'onlineSavingsMsg'
Learn More

Hexaphenylbenzene, 98+%

Product Code. 11360044
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11360044

Brand: Thermo Scientific Alfa Aesar L00446.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 992-04-1 | C42H30 | 534.702 g/mol

Hexaphenylbenzene was used to prepare the fluorescent nanorods used for the detection of trinitrotoulene (TNT).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexaphenylbenzene was used to prepare the fluorescent nanorods used for the detection of trinitrotoulene (TNT).

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 992-04-1
Molecular Formula C42H30
Molecular Weight (g/mol) 534.702
MDL Number MFCD00003057
InChI Key QBHWPVJPWQGYDS-UHFFFAOYSA-N
Synonym hexaphenylbenzene, benzene, hexaphenyl, 3',4',5',6'-tetraphenyl-o-terphenyl, acmc-20akri, m-terphenyl, 2',4',5',6'-tetraphenyl, ghl.pd_mitscher_leg0.890, hexaphenylbenzene 1g, 1,1':2',1-terphenyl, 3',4',5',6'-tetraphenyl, 1,2,3,4,5,6-hexaphenylbenzene
PubChem CID 70432
IUPAC Name 1,2,3,4,5,6-hexakis-phenylbenzene
SMILES C1=CC=C(C=C1)C2=C(C(=C(C(=C2C3=CC=CC=C3)C4=CC=CC=C4)C5=CC=CC=C5)C6=CC=CC=C6)C7=CC=CC=C7

Especificaciones

Melting Point >300°C
Quantity 1 g
Beilstein 1895903
Solubility Information Insoluble in water.
Formula Weight 534.7
Percent Purity ≥98%
Chemical Name or Material Hexaphenylbenzene

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado