missing translation for 'onlineSavingsMsg'
Learn More

(±)-Hexanoylcarnitine Chloride, TRC

Product Code. 30394061
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
2.50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394061 1 mg -
30395560 2.5 mg 2.50mg
30388827 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30394061 Supplier Toronto Research Chemicals Supplier No. TRCH2965651MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6920-35-0
Molecular Formula C13H26NO4+ • Cl-
Molecular Weight (g/mol) 260.353545
Synonym 1-Propanaminium, 3-carboxy-N,N,N-trimethyl-2-[(1-oxohexyl)oxy]-, chloride (1:1) (ACI), 1-Propanaminium, 3-carboxy-N,N,N-trimethyl-2-[(1-oxohexyl)oxy]-, chloride (9CI), 1-Propanaminium, 3-carboxy-N,N,N-trimethyl-2-[(1-oxohexyl)oxy]-, chloride, (±)- (ZCI), Ammonium, (3-carboxy-2-hydroxypropyl)trimethyl-, chloride, hexanoate, (±)- (8CI), Hexanoic acid, ester with (3-carboxy-2-hydroxypropyl)trimethylammonium chloride, (±)- (8CI), (3-Carboxy-2-hydroxypropyl)trimethylammonium chloride hexanoate, Hexanoyl carnitine hydrochloride, Hexanoyl-DL-carnitine hydrochloride, Hexanoylcarnitine chloride, 3-Carboxy-N,N,N-trimethyl-2-[(1-oxohexyl)oxy]-1-propanaminium chloride
SMILES O=C(OC(C[N+](C)(C)C)CC(O)=O)CCCCC.[Cl-]

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C13H25NO4.ClH/c1-5-6-7-8-13(17)18-11(9-12(15)16)10-14(2,3)4;/h11H,5-10H2,1-4H3;1H
Formula Weight 295.155
Chemical Name or Material (±)-Hexanoylcarnitine Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.