missing translation for 'onlineSavingsMsg'
Learn More

Hexanoyl-Lysine adduct Antibody (5E8), Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18426493 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18426493 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18426493 Supplier Novus Biologicals Supplier No. NBP259363JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Hexanoyl-Lysine adduct Monoclonal antibody specifically detects Hexanoyl-Lysine adduct in Nonspecies specific samples. It is validated for Western Blot, Flow Cytometry, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hexanoyl-Lysine adduct
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 5E8
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias HEL;HEL Adduct;Hexanoyl-Lys;Hexanoyl-Lys (HEL);Hexanoyl-Lys adduct
Host Species Mouse
Immunogen Synthetic Hexanoyl modified Keyhole Limpet Hemocyanin (KLH).
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.