missing translation for 'onlineSavingsMsg'
Learn More

Hexanoyl-Lysine adduct Antibody (5D9), mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18407792 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407792 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407792 Supplier Novus Biologicals Supplier No. NBP259362MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Hexanoyl-Lysine adduct Monoclonal antibody specifically detects Hexanoyl-Lysine adduct in Non-species specific samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hexanoyl-Lysine adduct
Applications Flow Cytometry
Classification Monoclonal
Clone 5D9
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias HEL;HEL Adduct;Hexanoyl-Lys;Hexanoyl-Lys (HEL);Hexanoyl-Lys adduct
Host Species Mouse
Immunogen Synthetic Hexanoyl modified Keyhole Limpet Hemocyanin (KLH).
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.