missing translation for 'onlineSavingsMsg'
Learn More

Hexanoyl-d3-L-carnitine Chloride, TRC

Product Code. 30406523
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30406523 1 mg 1mg
30393302 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30406523 Supplier Toronto Research Chemicals Supplier No. TRCH2944421MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C13 D3 H23 N O4 . Cl
Molecular Weight (g/mol) 298.821
Synonym (2R)-3-Carboxy-N,N,N-trimethyl-2-[(1-oxohexyl-d3)oxy]-1-propanaminium Chloride, (R)-3-Carboxy-N,N,N-trimethyl-2-[(1-oxohexyl-d3)oxy]-1-propanaminium Chloride, (R)-3-Carboxy-2-(hexan-d3-oyloxy)-N,N,N-trimethylpropan-1-aminium Chloride
IUPAC Name [(2R)-3-carboxy-2-(6,6,6-trideuteriohexanoyloxy)propyl]-trimethylazanium;chloride
SMILES [Cl-].[2H]C([2H])([2H])CCCCC(=O)O[C@H](CC(=O)O)C[N+](C)(C)C

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C13H25NO4.ClH/c1-5-6-7-8-13(17)18-11(9-12(15)16)10-14(2,3)4;/h11H,5-10H2,1-4H3;1H/t11-;/m1./s1/i1D3;
Formula Weight 298.174
Chemical Name or Material Hexanoyl-d3-L-carnitine Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.