missing translation for 'onlineSavingsMsg'
Learn More

Hexanoyl chloride, 97%

Product Code. p-7026800
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11486206 100 g 100g
11496206 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11486206 Supplier Thermo Scientific Alfa Aesar Supplier No. B23027.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 142-61-0 | C6H11ClO | 134.603 g/mol

Hexanoyl chloride has been used in the synthesis of (±)-7-butyl-6,8-dihydroxy-3-pentyl-3,4-dihydroisochromen-1-one, 14-methyl-1-octadecene. It is used as crystal liquid, pharmaceutical and organic synthesis material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexanoyl chloride has been used in the synthesis of (±)-7-butyl-6,8-dihydroxy-3-pentyl-3,4-dihydroisochromen-1-one, 14-methyl-1-octadecene. It is used as crystal liquid, pharmaceutical and organic synthesis material.

Solubility
Soluble in ether, chloroform.

Notes
Moisture sensitive. Store under inert gas. Incompatible with oxidizing agents, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-61-0
Molecular Formula C6H11ClO
Molecular Weight (g/mol) 134.603
MDL Number MFCD00000760
InChI Key YWGHUJQYGPDNKT-UHFFFAOYSA-N
Synonym caproyl chloride, n-caproyl chloride, hexanoylchloride, unii-i7ge8hg3b2, n-hexanoyl chloride, caproic acid chloride, hexanoic acid chloride, i7ge8hg3b2, caproylchloride, hexoylchlorid
PubChem CID 67340
IUPAC Name hexanoyl chloride
SMILES CCCCCC(=O)Cl

Specifications

Melting Point °C to 87°C
Density 0.963
Boiling Point 150°C to 153°C
Flash Point 50°C (122°F)
Assay Percent Range 0.98
Refractive Index 1.426
Quantity 100 g
UN Number UN2920
Sensitivity Moisture sensitive
Merck Index 14,1763
Solubility Information Soluble in ether,chloroform.
Formula Weight 134.61
Percent Purity 98%
Chemical Name or Material Hexanoyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.