missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexanoyl chloride, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
Hexanoyl chloride has been used in the synthesis of (±)-7-butyl-6,8-dihydroxy-3-pentyl-3,4-dihydroisochromen-1-one, 14-methyl-1-octadecene. It is used as crystal liquid, pharmaceutical and organic synthesis material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Hexanoyl chloride has been used in the synthesis of (±)-7-butyl-6,8-dihydroxy-3-pentyl-3,4-dihydroisochromen-1-one, 14-methyl-1-octadecene. It is used as crystal liquid, pharmaceutical and organic synthesis material.
Solubility
Soluble in ether, chloroform.
Notes
Moisture sensitive. Store under inert gas. Incompatible with oxidizing agents, water/ moisture.
Hexanoyl chloride has been used in the synthesis of (±)-7-butyl-6,8-dihydroxy-3-pentyl-3,4-dihydroisochromen-1-one, 14-methyl-1-octadecene. It is used as crystal liquid, pharmaceutical and organic synthesis material.
Solubility
Soluble in ether, chloroform.
Notes
Moisture sensitive. Store under inert gas. Incompatible with oxidizing agents, water/ moisture.
Specifications
Melting Point | °C to 87°C |
Density | 0.963 |
Boiling Point | 150°C to 153°C |
Flash Point | 50°C (122°F) |
Assay Percent Range | 0.98 |
Refractive Index | 1.426 |
Quantity | 100 g |
UN Number | UN2920 |
Sensitivity | Moisture sensitive |
Merck Index | 14,1763 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'