missing translation for 'onlineSavingsMsg'
Learn More

Hexanophenone, 98%

Product Code. 10214970 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10214970 25 g Glass bottle 25g
10361614 5 g Glass bottle 5g
13468328 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10214970 Supplier Thermo Scientific Acros Supplier No. 120720250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 942-92-7 | C12H16O | 176.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 942-92-7
Molecular Formula C12H16O
Molecular Weight (g/mol) 176.26
MDL Number MFCD00009512
InChI Key MAHPVQDVMLWUAG-UHFFFAOYSA-N
Synonym hexanophenone, caprophenone, 1-hexanone, 1-phenyl, 1-phenyl-1-hexanone, n-hexanophenone, amyl phenyl ketone, hexaphenone, pentyl phenyl ketone, 4-ethylbutyrophenone, n-amylphenyl ketone
PubChem CID 70337
IUPAC Name 1-phenylhexan-1-one
SMILES CCCCCC(=O)C1=CC=CC=C1

Specifications

Melting Point 25.0°C to 26.0°C
Color Yellow
Density 0.9500g/mL
Boiling Point 265.1°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C6H5CO(CH2)4CH3
Refractive Index 1.5105
Quantity 25 g
Beilstein 07,II,257
Specific Gravity 0.95
Formula Weight 176.26
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material Hexanophenone, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.