missing translation for 'onlineSavingsMsg'
Learn More

Hexanoic anhydride, 98%

Product Code. 11470198
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11470198 250 g 250g
11460198 50 g 50g
11450198 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11470198 Supplier Thermo Scientific Alfa Aesar Supplier No. B23756.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2051-49-2 | C12H22O3 | 214.305 g/mol

It is used as a reactant in the synthesis of acremomannolipin A. It is used in green synthesis of esters of acyclovir (acyclovir prodrugs), in the preparation of hexanoyl-modified chitosan nanoparticles, chitosan-based polymeric surfactants via N-acylation of chitosans.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a reactant in the synthesis of acremomannolipin A. It is used in green synthesis of esters of acyclovir (acyclovir prodrugs), in the preparation of hexanoyl-modified chitosan nanoparticles, chitosan-based polymeric surfactants via N-acylation of chitosans.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store in cool, dry conditions in well sealed containers. Store away from oxidizing agents, bases, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2051-49-2
Molecular Formula C12H22O3
Molecular Weight (g/mol) 214.305
MDL Number MFCD00009509
InChI Key PKHMTIRCAFTBDS-UHFFFAOYSA-N
Synonym hexanoic anhydride, caproic anhydride, hexanoic acid, anhydride, n-hexanoic anhydride, hexanoic acid, 1,1'-anhydride, unii-gg70y8ltby, n-caproic anhydride, caproic acid anhydride, capronic acid anhydride, n-hexanoic acid anhydride
PubChem CID 74918
IUPAC Name hexanoyl hexanoate
SMILES CCCCCC(=O)OC(=O)CCCCC

Specifications

Melting Point -40°C
Density 0.928
Boiling Point 246°C to 248°C
Flash Point 128°C (262°F)
Odor Unpleasant
Linear Formula [CH3(CH2)4CO]2O
Refractive Index 1.429
Quantity 250 g
UN Number UN3265
Beilstein 1776561
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 214.31
Percent Purity 98%
Chemical Name or Material Hexanoic anhydride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.