missing translation for 'onlineSavingsMsg'
Learn More

Hexanoic anhydride, 97%

Product Code. 10032340 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10032340 250 g Glass bottle 250g
10636582 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10032340 Supplier Thermo Scientific Acros Supplier No. 186752500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2051-49-2 | C12H22O3 | 214.3 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2051-49-2
Molecular Formula C12H22O3
Molecular Weight (g/mol) 214.3
MDL Number MFCD00009509
InChI Key PKHMTIRCAFTBDS-UHFFFAOYSA-N
Synonym hexanoic anhydride, caproic anhydride, hexanoic acid, anhydride, n-hexanoic anhydride, hexanoic acid, 1,1'-anhydride, unii-gg70y8ltby, n-caproic anhydride, caproic acid anhydride, capronic acid anhydride, n-hexanoic acid anhydride
PubChem CID 74918
IUPAC Name hexanoyl hexanoate
SMILES CCCCCC(=O)OC(=O)CCCCC

Specifications

Melting Point -40°C
Color Colorless to Yellow
Density 0.9260g/mL
Boiling Point 246°C to 248°C
Flash Point 130°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula [CH3(CH2)4CO]2O
Refractive Index 1.4270 to 1.429
Quantity 250 g
Beilstein 02, 324
Specific Gravity 0.926
Solubility Information Solubility in water: 72g/L. Other solubilities: soluble in ethanol and ether
Formula Weight 214.3
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Hexanoic anhydride, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Hexanoic anhydride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.