missing translation for 'onlineSavingsMsg'
Learn More

Hexanoic acid 10 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30325633
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30325633 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30325633 Supplier Dr. Ehrenstorfer Supplier No. DREA14196000AL10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Caproic Acid
CAS 142-62-1
Analyte or Component Names Caproic Acid
Shipping Condition Room Temperature
Molecular Formula C6 H12 O2
Concentration 10 μg/mL
Certifications/Compliance ISO 17034
Synonym Hexanoic acid, Hexanoic acid (8CI, 9CI, ACI), 1-Hexanoic acid, 1-Pentanecarboxylic acid, Butylacetic acid, Capronic acid, Hexoic acid, Hexylic acid, n-Caproic acid, n-Hexanoic acid, n-Hexoic acid, n-Hexylic acid, NAA 60, NSC 35598, NSC 8266, Pentylformic acid, C6:0
InChI Formula InChI=1S/C6H12O2/c1-2-3-4-5-6(7)8/h2-5H2,1H3,(H,7,8)
SMILES CCCCCC(=O)O
IUPAC Name hexanoic acid
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 116.16
Quantity 1 mL
Formula Weight 116.0837 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.