missing translation for 'onlineSavingsMsg'
Learn More

Hexanoic-6,6,6-d3 Acid, TRC

Product Code. 30406515
Change view
Click to view available options
Quantity:
5 mg
50 mg
100 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30406515 100 mg -
30399866 50 mg 50mg
30400556 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30406515 Supplier Toronto Research Chemicals Supplier No. TRCH292147100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55320-69-9
Molecular Formula C6 D3 H9 O2
Molecular Weight (g/mol) 119.18
Synonym 1-Hexanoic-d3 Acid, 1-Pentane-d3-carboxylic Acid, Butyl-d3-acetic Acid, Caproic-d3 Acid, Capronic-d3 Acid, Hexoic-d3 Acid, Hexylic-d3 Acid, NSC 35598-d3, NSC 8266-d3, Pentylformic-d3 Acid, n-Caproic-d3 Acid, n-Hexanoic-d3 Acid, n-Hexoic-d3 Acid, n-Hexylic-d3 Acid, Hexanoic-6,6,6-d3 acid (9CI), Hexanoic acid-d3, Hexanoic-ω-d3 acid, Caproic-ω-d3 acid
IUPAC Name 6,6,6-trideuteriohexanoic acid
SMILES [2H]C([2H])([2H])CCCCC(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C6H12O2/c1-2-3-4-5-6(7)8/h2-5H2,1H3,(H,7,8)/i1D3
Formula Weight 119.1026
Chemical Name or Material Hexanoic-6,6,6-d3 Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.