missing translation for 'onlineSavingsMsg'
Learn More

Hexanes, mixed isomers, 98+%

Product Code. 11367976
Change view
Click to view available options
Quantity:
500 mL
1000 mL
2500 mL
Unit Size:
1000mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11367976 500 mL 500mL
11459830 1000 mL 1000mL
11469830 2500 mL 2500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 11367976 Supplier Thermo Scientific Alfa Aesar Supplier No. L13233.AP
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 92112-69-1 | C6H14 | 86.18 g/mol

Hexane mixed isomers are used in the laboratory to extract oil and grease contaminants from water and soil for analysis. As a non-polar solvent, it is used in chromatography. It plays an important role in the preparation of glues for shoes, leather products, and roofing. It is also used for reactions that involve very strong bases such as the preparation of Grignard reagents. Also involved in the preparation of organolithiums compounds, namely butyl lithium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexane mixed isomers are used in the laboratory to extract oil and grease contaminants from water and soil for analysis. As a non-polar solvent, it is used in chromatography. It plays an important role in the preparation of glues for shoes, leather products, and roofing. It is also used for reactions that involve very strong bases such as the preparation of Grignard reagents. Also involved in the preparation of organolithiums compounds, namely butyl lithium.

Solubility
Miscible with alcohol, chloroform, acetone and ether. Immiscible with water.

Notes
Moisture sensitive. Incompatible with oxidizing agents. Keep away from sources of ignition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92112-69-1
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD00009520
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Melting Point -95°C
Density 0.67
Boiling Point 65°C to 70°C
Flash Point <−23°C (−9°F)
Refractive Index 1.3795
Quantity 500 mL
UN Number UN1208
Solubility Information Miscible with alcohol,chloroform,acetone and ether. Immiscible with water.
Formula Weight 86.18
Percent Purity ≥98%
Chemical Name or Material Hexanes, mixed isomers
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.