missing translation for 'onlineSavingsMsg'
Learn More

Hexanes, 95+%, for spectroscopy, mixture of isomers

Product Code. 10327970 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
2.5 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10327970 2.5 L Glass bottle 2.5 L
10616572 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 10327970 Supplier Thermo Scientific Acros Supplier No. 168570025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 92112-69-1 | C6H14 | 86.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by a SFI certified supplier.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hexanes
Acidity 0.00015 meq/g max.
CAS 92112-69-1, 110-54-3
Absorbance (1 cm cell vs water), 0.005 max. at 240nm, 0.005 max. at 250nm, 0.01 max. at 230nm, 0.025 max. at 220nm, 0.1 max. at 210nm, 0.3 max. at 200nm
Melting Point -95°C
Density 0.6590g/mL
Boiling Point 69°C
Flash Point -22°C
Assay Percent Range (95% n-hexane approximately) (GC)
Packaging Glass bottle
Molecular Formula C6H14
Linear Formula CH3(CH2)4CH3
Refractive Index 1.3748 to 1.381
MDL Number MFCD00009520
Quantity 2.5 L
Beilstein 01, 142
Residue after Evaporation 0.0005% max.
Merck Index 14, 4694
Specific Gravity 0.659
Synonym Hex
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,acetone,ether and chloroform
Molecular Weight (g/mol) 86.18
Viscosity 0.31 mPa.s (20°C)
Formula Weight 86.18
Vapor Pressure 160mbar at 20°C
Percent Purity 95+%
Grade Spectroscopy
Physical Form Liquid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.