missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexanedioic Acid Mono(4-hydroxybutyl) Ester, TRC
Specifications
Specifications
| Chemical Name or Material | Hexanedioic Acid Mono(4-hydroxybutyl) Ester |
| CAS | 116789-12-9 |
| Molecular Formula | C10 H18 O5 |
| Quantity | 100 mg |
| Synonym | Hexanedioic Acid 1-(4-Hydroxybutyl) Ester |
| InChI Formula | InChI=1S/C10H18O5/c11-7-3-4-8-15-10(14)6-2-1-5-9(12)13/h11H,1-8H2,(H,12,13) |
| SMILES | OCCCCOC(=O)CCCCC(=O)O |
| IUPAC Name | 6-(4-hydroxybutoxy)-6-oxohexanoic acid |
| Molecular Weight (g/mol) | 218.247 |
| Formula Weight | 218.115 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?