missing translation for 'onlineSavingsMsg'
Learn More

Hexane, mixture of isomers, Honeywell™

Product Code. 15664760
Change view
Click to view available options
:
Quantity: 2.5 L; Packaging: Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15664760 Quantity: 2.5 L; Packaging: Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 15664760 Supplier Honeywell Riedel-de Haën Supplier No. 349942.5L
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Chromasolv™ For HPLC

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hexane, mixture of isomers
Melting Point -154°C
CAS 110-54-3, 68476-44-8, 68956-52-5, 92112-69-1
Color Colorless
Quantity 2.5 L
Packaging Glass Bottle
Boiling Point 48°C to 65°C (1013 hPa)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.