Product Code. 15646120
Click to view available options
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Packaging
unitSize
15646120
1 L
Glass Bottle
1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code.
15646120
Supplier
Honeywell-Riedel-de Haen
Supplier No.
6505441L
This item is currently unavailable or has been discontinued. View the product page for possible alternatives.
View alternative products
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Specifications
CAS
110-54-3
Density
0.672g/mL at 25°C
Quantity
1 L
Explosion Limit
0.075
Flash Point
−23°C (−9.4°F)
Ignition Point
437°F
Packaging
Glass Bottle
Molecular Formula
C6H14
MDL Number
MFCD02179311
UN Number
UN1208
Beilstein
1730733
Synonym
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
InChI Key
VLKZOEOYAKHREP-UHFFFAOYSA-N
SMILES
CCCCCC
IUPAC Name
hexane
Molecular Weight (g/mol)
86.18
PubChem CID
8058
ChEBI
CHEBI:29021
Formula Weight
86.18g/mol
Percent Purity
≥98.5%
Grade
HPLC Plus
Chemical Name or Material
Hexane, mixture of isomers
Show More
Show Less
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?