missing translation for 'onlineSavingsMsg'
Learn More

Hexane, Honeywell

Product Code. 15673760
Change view
Click to view available options
Quantity:
1 L
2.5 L
25 L
Unit Size:
1 L
2.5 L
25 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15673760 25 L 25 L
15653760 1 L 1 L
15663760 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 15673760 Supplier Honeywell Riedel-de Haën Supplier No. 3229325L
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Density 0.659g/mL at 25°C
Quantity 25 L
Explosion Limit 0.077
Flash Point −26°C (−14.8°F)
Ignition Point 453°F
Linear Formula CH3(CH2)4CH3
Vapor Pressure 5.2psi (37.7°C), 132mmHg (20°C), 256mmHg (37.7°C)
UN Number 1208
Beilstein 1730733
Formula Weight 86.18g/mol
Percent Purity ≥99% (GC)
Grade ACS/puriss. p.a.
Chemical Name or Material Hexane, Reag. Ph. Eur.
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hexane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.