Product Code. 15612850
Click to view available options
Quantity:
1 L
2.5 L
6 x 1 L
Unit Size:
1 L
2.5 L
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Packaging
unitSize
15612850
1 L
Glass Bottle
1 L
15622850
2.5 L
Glass Bottle
2.5 L
15642850
6 x 1 L
Glass Bottle
Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code.
15612850
Supplier
Honeywell-Chromasolv
Supplier No.
2705041L
This item is currently unavailable or has been discontinued. View the product page for possible alternatives.
View alternative products
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
CHROMASOLV™, for HPLC,≥95%
For HPLC and organic synthesis High-purity, multipurpose solvents High lot-to-lot consistency
Chemical Identifiers
CAS
110-54-3
Molecular Formula
C6H14
Molecular Weight (g/mol)
86.18
MDL Number
MFCD02179311
InChI Key
VLKZOEOYAKHREP-UHFFFAOYSA-N
PubChem CID
8058
ChEBI
CHEBI:29021
IUPAC Name
hexane
SMILES
CCCCCC
Specifications
Melting Point
-95°C (literature)
Absorbance
max. 0.02 at 240 nm, max. 0.01 at 250 nm, max. 0.01 at 400 nm, max. 1.0 at 195 nm, max. 0.30 at 210 nm, max. 0.10 at 220 nm, max. 0.05 at 230 nm
CAS Min %
≥90.0000%
CAS Max %
≤100.0000%
Density
0.659 g/mL at 25 °C(lit.)
Quantity
1 L
Flash Point
−26°C
Packaging
Glass Bottle
Linear Formula
CH3(CH2)4CH3
UN Number
UN1208
Beilstein
1730733
Synonym
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
Boiling Point
66°C to 69°C
Formula Weight
86.18
Percent Purity
>95%
Grade
HPLC
Chemical Name or Material
Hexane, CHROMASOLV™, for HPLC, ≥95%
Show More
Show Less
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?