missing translation for 'onlineSavingsMsg'
Learn More

Solstice Hexane CHROMASOLV™

Product Code. 15905510
Change view
Click to view available options
Quantity:
7 L
18 L
45 L
200 L
Unit Size:
18 L
200 L
45 L
7 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15905510 7 L 7 L
15995660 18 L 18 L
15995500 45 L 45 L
16653954 200 L 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 15905510 Supplier Solstice Supplier No. 348597L
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Hexane CHROMASOLV™ for HPLC and organic synthesis, ≥97.0% (GC).

  • For HPLC and organic synthesis.
  • High-purity, multipurpose solvents.
  • High lot-to-lot consistency.

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-Hexane
IUPAC Name hexane
SMILES CCCCCC

Specifications

pH Not applicable
Melting Point -95°C
Color Colorless
CAS Max % 100.00%
Density 0.660 to 0.668 g/cm3 (20°C)
Quantity 7 L
Flash Point 7.4 %(V)
Packaging Returnable Stainless Steel Can
Physical Form Liquid
Product Line CHROMASOLV
Linear Formula CH3(CH2)4CH3
Vapor Pressure 160 hPa (20°C)
Viscosity 0.31 mPa.s (20°C)
UN Number UN1208
Beilstein 1730733
Concentration 100.00%
Boiling Point 66°C to 69°C (1.013 hPa)
Percent Purity ≥97.00%
Grade HPLC
Chemical Name or Material Hexane
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hexane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.