missing translation for 'onlineSavingsMsg'
Learn More

Hexane, 99% n-Hexane, BAKER ANALYZED, J.T. Baker™

Product Code. 10529221 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10529221 1 L Glass 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 10529221 Supplier JT Baker Supplier No. 8205.1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

High quality chemical of GC-Spectrophotometric quality that exceeds ACS specifications for laboratory and specialized industrial use.

  • Does not interfere with the measurement at the specific wavelength being measured
  • Exceeds the requirements set forth by the American Chemical Society (ACS)
  • Label provides an actual Lot Analysis as a guarantee of purity and consistency
  • Analytical methods used for these determinations are based on standard test procedures described in technical publications
  • Improvements developed by J.T. Baker are employed to achieve a higher degree of reliability

Specifications

Melting Point -95°C
Absorbance Max. 1.00 at 210nm, Max. 0.20 at 220nm, Max. 0.10 at 230nm, Max. 0.04 at 240nm, Max. 0.02 (at 250nm) / 0.01 (at 280-400nm)
CAS 110-54-3
Color Colorless
Density 0.657 g/mL at 25°C
Quantity 1 L
Explosion Limit 1.2 - 7.4 Vol %
Flash Point −26°C
Ignition Point 240°C
Packaging Glass
Physical Form Liquid
Residue after Evaporation Max. 5 ppm
Molecular Formula C6H14
Linear Formula CH3(CH2)4CH3
MDL Number MFCD02179311
Viscosity 0.326 mPas (dynamic), 0.50mm2/s (kinematic)
UN Number 1208
Beilstein 1730733
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
Boiling Point 69°C
Solubility Information 0.1g/L in water
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
SMILES CCCCCC
IUPAC Name hexane
Molecular Weight (g/mol) 86.18
PubChem CID 8058
ChEBI CHEBI:29021
Formula Weight 86.18g/mol
Grade Exceeds ACS specifications, GC-Spectrophotometric quality
Chemical Name or Material n-Hexane
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.