missing translation for 'onlineSavingsMsg'
Learn More

Hexane-1,6-bis(tri-n-butylammonium) dihydroxide, 20% w/w aq. soln.

Product Code. 11398878
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
Unit Size:
1 L
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11398878 250 mL 250mL
11378878 50 mL 50mL
11388878 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11398878 Supplier Thermo Scientific Alfa Aesar Supplier No. 041725.AK
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 69762-88-5 | C30H68N2O2 | 488.886 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69762-88-5
Molecular Formula C30H68N2O2
Molecular Weight (g/mol) 488.886
MDL Number MFCD00216633
InChI Key ALYGOOWRCDZDTJ-UHFFFAOYSA-L
Synonym tributyl-6-tributylazaniumyl hexyl azanium,dihydroxide, n1,n1,n1,n6,n6,n6-hexabutylhexane-1,6-diaminium hydroxide, tributyl 6-tributylammonio hexyl azanium dihydroxide, n,n,n,n,n,n-hexabutylhexamethylenediammonium dihydroxide solution, hexane-1,6-bis tributylammonium dihydroxide w/w aqueous solution, n,n inverted exclamation marka-hexamethylenebis tributylammonium hydroxide, n∼1∼,n∼1∼,n∼1∼,n∼6∼,n∼6∼,n∼6∼-hexabutylhexane-1,6-bis aminium dihydroxide, n,n,n,n inverted exclamation marka,n inverted exclamation marka,n inverted exclamation marka-hexabutylhexamethylenediammonium dihydroxide solution
PubChem CID 15376281
IUPAC Name tributyl-[6-(tributylazaniumyl)hexyl]azanium;dihydroxide
SMILES CCCC[N+](CCCC)(CCCC)CCCCCC[N+](CCCC)(CCCC)CCCC.[OH-].[OH-]

Specifications

Quantity 250 mL
UN Number UN3267
Formula Weight 488.87
Concentration or Composition (by Analyte or Components) 20% w/w aq. soln.
Physical Form Liquid
Chemical Name or Material Hexane-1,6-bis(tri-n-butylammonium) dihydroxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.