missing translation for 'onlineSavingsMsg'
Learn More

Hexanamide, 98%, Thermo Scientific™

Product Code. 10791632
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 628-02-4
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.18
MDL Number MFCD00008046
InChI Key ALBYIUDWACNRRB-UHFFFAOYSA-N
Synonym capronamide, hexamide, hexylamide, n-caproamide, caproamide, n-hexanamide, hexamide van, hexanoamide, n-capronamide, n-caproic amide
PubChem CID 12332
IUPAC Name hexanamide
SMILES CCCCCC(=O)N

Specifications

Melting Point 100.0°C to 102.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3(CH2)4CONH2
Quantity 1 g
Formula Weight 115.18
Percent Purity 98%
Physical Form Flakes
Chemical Name or Material Hexanamide, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Hexanamide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'