missing translation for 'onlineSavingsMsg'
Learn More

Hexanal, TRC

Product Code. 30406001
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30406001 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30406001 Supplier Toronto Research Chemicals Supplier No. TRCH29213050G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 66-25-1
Molecular Formula C6 H12 O
Quantity 50 g
Synonym Hexanal, Caproaldehyde, Caproic aldehyde, Capronaldehyde, Hexaldehyde, Hexanaldehyde, Hexylaldehyde, NSC 2596, n-Caproaldehyde, n-Capronaldehyde, n-Hexanal, n-Hexylaldehyde
InChI Formula InChI=1S/C6H12O/c1-2-3-4-5-6-7/h6H,2-5H2,1H3
SMILES CCCCCC=O
IUPAC Name hexanal
Molecular Weight (g/mol) 100.16
Formula Weight 100.0888
Chemical Name or Material Caproic aldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.