missing translation for 'onlineSavingsMsg'
Learn More

Hexanal dimethyl acetal, 98%, Thermo Scientific™

Product Code. 11493098
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11493098 25 g 25g
11403108 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11493098 Supplier Thermo Scientific Alfa Aesar Supplier No. A18824.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1599-47-9
Molecular Formula C8H18O2
Molecular Weight (g/mol) 146.23
MDL Number MFCD00036508
InChI Key ZQUJUCJLDXTKKW-UHFFFAOYSA-N
PubChem CID 74137
IUPAC Name 1,1-dimethoxyhexane
SMILES CCCCCC(OC)OC

Specifications

Density 0.847
Boiling Point 163°C to 164°C
Flash Point 54°C (130°F)
Linear Formula CH3(CH2)4CH(OCH3)2
Refractive Index 1.405
Quantity 25 g
UN Number UN1993
Synonym hexanal dimethyl acetal, hexane, 1,1-dimethoxy, aldehyde c-6 dimethyl acetal, acmc-20akrj, 1,1-dimethoxy-hexane, hexane,1,1-dimethoxy, hexaldehyde dimethyl acetal, 1,1-dimethoxyhexane
Formula Weight 146.23
Percent Purity 98%
Chemical Name or Material Hexanal dimethyl acetal
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.