missing translation for 'onlineSavingsMsg'
Learn More

Hexanal, 98%

Product Code. 11457727
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11457727 500 g 500g
11447727 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11457727 Supplier Thermo Scientific Alfa Aesar Supplier No. A16265.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 66-25-1 | C6H12O | 100.161 g/mol

Hexanal is used in the flavor industry to prepare fruity flavors. It is utilized as a flavoring agent in the food industry. It is also used in Witting and aldol reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexanal is used in the flavor industry to prepare fruity flavors. It is utilized as a flavoring agent in the food industry. It is also used in Witting and aldol reactions.

Solubility
Miscible with ethanol, ethyl ether, acetone, benzene and water.

Notes
Air sensitive. Incompatible with oxidizing agents, strong acids, strong bases and strong reducing agents. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66-25-1
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00007027
InChI Key JARKCYVAAOWBJS-UHFFFAOYSA-N
Synonym caproaldehyde, hexaldehyde, caproic aldehyde, capronaldehyde, 1-hexanal, n-hexanal, hexanaldehyde, n-caproaldehyde, hexylaldehyde, aldehyde c-6
PubChem CID 6184
IUPAC Name hexanal
SMILES CCCCCC=O

Specifications

Melting Point -56°C
Density 0.834
Boiling Point 130°C to 131°C
Flash Point 25°C (77°F)
Assay Percent Range 98%
Refractive Index 1.405
Quantity 500 g
UN Number UN1207
Beilstein 506198
Sensitivity Air Sensitive
Merck Index 14,1760
Solubility Information Miscible with ethanol,ethyl ether,acetone,benzene and water.
Formula Weight 100.16
Percent Purity 98%
Chemical Name or Material Hexanal
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hexanal
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Hexaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.